Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Dextromethorphan Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Dextromethorphan Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 °C
mobile phase [A] methanol; [B] 20 mM ammonium acetate, pH 4.1; (90:10, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antitussives, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC