Attention:
As we update our core systems, key features of the website including ordering and real-time price and availability, may be unavailable from 8:00 PM CDT Friday, June 21st until 9:00 AM CDT Saturday morning June 22nd.
We apologize for any inconvenience this may cause.
HPLC Analysis of Diet Soda Additives on Ascentis® Express RP-Amide
HPLC Analysis of Diet Soda Additives on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 3 cm x 4.6 mm I.D., 2.7 µm particles (53921-U)
column temp. 40 °C
mobile phase [A] 20 mM ammonium acetate, pH 5.6 with acetic acid; [B] water; [C] acetonitrile
gradient 20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
flow rate 3.0 mL/min
pressure 3336 psi (230 bar)
sample Diet Soda 100 - 500 µg/mL in buffer
injection 1 µL
detector UV, 214 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola.
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Soft Drinks
Featured Industry Food and Beverages