Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Diet Soda Spiked with Neotame on Ascentis® Express C18
HPLC Analysis of Diet Soda Spiked with Neotame on Ascentis® Express C18
Conditions
column Ascentis Express C18, 3 cm x 4.6 mm I.D., 2.7 µm particles (53818-U)
column temp. 40 °C
mobile phase [A] 100 mM ammonium acetate, pH 5.6 (titrated with acetic acid); [B] water; [C] acetonitrile
gradient 20% A constant; 75 to 20% B, 5 to 60% C in 1 min; held at 20% B, 60% C for 0.1 min
flow rate 3 mL/min
pressure 3191 psi (220 bar)
sample neotame 350 µg/mL in diet cola
injection 1 µL
detector UV, 214 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of common drink additives used in diet soda.
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Soft Drinks
Featured Industry Food and Beverages