Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Disaccharides Turanose, Lactulose, and Melibiose on Chromolith® NH2
HPLC Analysis of Disaccharides Turanose, Lactulose, and Melibiose on Chromolith® NH2
Conditions
column Chromolith NH2, 10 cm x 4.6 mm I.D., 2 µm particles (1.52028.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] water; (85:15, A:B)
flow rate 4 mL/min
pressure 1102 psi (76 bar)
sample turanose 7.7 mg/mL, lactulose 8.0 mg/mL, melibiose 10.6 mg/mL in mobile phase
injection 10 µL
detector UV, 190 nm
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany