HPLC Analysis of Disaccharides - Turanose, Lactulose, Melibiose on Chromolith(R) NH2 100 - 4.6mm
HPLC Analysis of Disaccharides - Turanose, Lactulose, Melibiose on Chromolith® NH2 100 - 4.6mm
Conditions
column Chromolith® NH2, 100 x 4.6 mm (1.52028.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] water; (85:15, A:B)
flow rate 4.0 mL/min
pressure 1102 psi (76 bar)
sample Turanose 7.7mg/mL, Lactulose 8.0 mg/mL, Melibiose 10.6mg/mL in mobile phase
injection 10 μL
detector 190 nm
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany