Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Diuretic Drugs on Ascentis® C18
HPLC Analysis of Diuretic Drugs on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] 10 mM monobasic potassium phosphate (pH 3.0 with phosphoric acid); [B] acetonitrile; (A:B, 40:60)
flow rate 1 mL/min
sample 10 µg/mL each in mobile phase
injection 10 µL
detector UV, 234 nm
Description
Categories Analytical Chromatography, Diuretics