Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Dopamine Receptor Antagonists on Ascentis® Phenyl
HPLC Analysis of Dopamine Receptor Antagonists on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 °C
mobile phase [A] 50 mM ammonium formate (pH 3.0 with formic acid); [B] acetonitrile; (45:55, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL in 50:50, water:methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, neurotransmitters, catecholamines, Hormones, hormones, antidepressants
Featured Industry Clinical
Life Science and Biopharma