Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Drugs of Abuse on Ascentis® Phenyl
HPLC Analysis of Drugs of Abuse on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 °C
mobile phase [A] water with 0.1% ammonium acetate, pH 6.6, unadjusted; [B] acetonitrile with 0.1% ammonium acetate
gradient 25 to 40% B in 7 min; to 25% B in 1 min
flow rate 1.0 mL/min
sample as indicated in mobile phase A
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Analgesics
Featured Industry Forensics and Toxicology