DART(R)-MS Analysis of Drugs of Abuse in Human Urine Using C18 SPME LC (BioSPME) Tips
DART®-MS Analysis of Drugs of Abuse in Human Urine Using C18 SPME LC (BioSPME) Tips
Conditions
sample/matrix human urine
desorption process Direct MS
extraction 60 min with 1 mL sample
SPME fiber SPME LC Tips C18 (57234-U)
detector MS, Full scan 100-500 amu
Description
Analysis Note Economical and user-friendly sample prep method for DART-MS detection and identification of drugs of abuse in urine.
Featured Industry Forensics and Toxicology
Legal Information DART is a registered trademark of JEOL USA, Inc.
suitability application for SPME, application for HPLC