Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Elastase on Discovery® BIO PolyMA-SCX, Ion Exchange
HPLC Analysis of Elastase on Discovery® BIO PolyMA-SCX, Ion Exchange
Conditions
column Discovery BIO PolyMA-SCX, 5cm x 4.6mm, 5mum particles (59601-U)
column temp. 25 °C
mobile phase [A] 20 mM sodium phosphate, pH 7.0; [B] 20 mM sodium phosphate, 0.5 M NaCl, pH 7.0
gradient 0 to 100% B in 30 min. (linear)
flow rate 0.5 mL/min
injection 10 µL
detector UV, 280 nm
Description
Analysis Note Discovery BIO PolyMA-SCX and PolyMA-WAX columns give sharp, efficient peaks for a wide variety of proteins.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC