Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ephedrine Compounds on Ascentis® Express HILIC
HPLC Analysis of Ephedrine Compounds on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 35 °C
mobile phase [A] acetonitrile; [B] 40 mM ammonium acetate in water; (90:10, A:B)
flow rate 0.4 mL
sample 150 µg/mL each in mobile phase
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Decongestants, Bronchodilators, StiµLants, Antihistamines