HPLC Analysis of Epoxy resin, Epikote(R) 1001 on TSKgel(R) G3000HXL
HPLC Analysis of Epoxy resin, Epikote® 1001 on TSKgel® G3000HXL
Conditions
column TSKgel G3000HXL, 30 cm × 7.8 mm I.D., 6 μm particles (816136)
mobile phase tetrahydrofuran
flow rate 1 mL/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Volatiles
Featured Industry Chemicals and Industrial Polymers
Legal Information Epikote is a registered trademark of Hexion, Inc., TSKgel is a registered trademark of Tosoh Corporation