Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Erectile Dysfunction Drugs on Ascentis® Express RP-Amide
HPLC Analysis of Erectile Dysfunction Drugs on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 3.0 mm I.D., 5 µm particles (50753-U)
column temp. 35 °C
mobile phase [A] water with 0.1% formic acid; [B] acetonitrile with 0.1% formic acid;
gradient 5% B held for 0.5 min; to 100% B in 9.5 min; 100% B held for 2 min
flow rate 0.4 mL/min
pressure 870 psi (60 bar)
sample 20 µg/mL in water:methanol (85:15)
injection 2 µL
detector UV, 250 nm
Description
Categories Analytical Chromatography, ED Drugs