HPLC Analysis of Ergosterol in Dust on Chromolith(R) Performance 100-2mm
HPLC Analysis of Ergosterol in Dust on Chromolith® Performance 100-2mm
Conditions
column Chromolith® Performance, 100 x 2 mm (1.52006.0001)
column temp. 23 °C
mobile phase [A] water; [B] methanol; (2:98, A:B)
flow rate 190 μL/min
pressure 595 psi 41 Bar
sample Standard: Ergosterine 1mg/mL (Diluent: Standard: Methanol)
injection 1.0 μL
detector 282 nm cell: 1.4 μL
Description
Featured Industry Industrial Hygiene
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany