Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ergosterol in Vacuum Cleaner Dust on Chromolith(R) Performance 100-2mm
HPLC Analysis of Ergosterol in Vacuum Cleaner Dust on Chromolith® Performance 100-2mm
Conditions
sample preparation See "Analysis Note."
column Chromolith® Performance, 100 x 2 mm (1.52006.0001)
column temp. 23 °C
mobile phase [A] methanol; [B] Water; (98:2, A:B)
flow rate 190 μL/min
pressure 595 psi 41 Bar
sample Standard: Ergosterol; Matrix: Dust of a vacuum cleaner
injection 1 μL
detector 282 nm cell: 1.4 μL
Description
Analysis Note Sample preparation: The dust of a vacuum cleaner was sieved through a normal kitchen sieve. 500 mg of the sieved dust was suspended with 5.4 mL potassium hydroxide (10%) in methanol and stirred over 2h by 80°C.After cooling down, the suspension  was centrifuged. 4mL of the obtained solution was filtered through a 0.45 μm  membrane filter and washed with 2mL water. The combined solutions were then extracted two times with 2 mL n-hexane. The n-hexane was evaporated with nitrogen gas and the residue was resolved in 1 mL methanol. The  final solution  were  spiked with 100 μL of Ergosterol  (1 mg/mL) standard solution .
Featured Industry Industrial Hygiene
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany