Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Estrogens on Ascentis® Express Biphenyl
HPLC Analysis of Estrogens on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10 cm x 2.1 mm I.D., 2.7 µm particles (64065-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile, (60:40, A:B)
flow rate 0.4 mL/min
pressure 2900 psi (200 bar)
sample 60 µg/mL each in 50:50, water:organic solvent
injection 2 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Steroids
Featured Industry Clinical