Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Fab/Fc mAb Antibody Fragments on Proteomix® WCX NP1.7
HPLC Analysis of Fab/Fc mAb Antibody Fragments on Proteomix® WCX NP1.7
Conditions
column Proteomix WCX-NP1.7, 3 cm x 4.6 mm I.D., 1.7 µm particles (Z777179)
mobile phase (A) 20 mM sodium acetate, pH 5.15; (B) A + 1 M LiCl
gradient 12% to 40% B in 6.7 min; to 100% B in 0.1 min; held for 2 min
flow rate (1.5 mL/min)
pressure 3335-3916 psi (230 - 270 bar)
sample 1 mg/mL papain digested MAb 321
injection 25 µL
detector UV 280 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies