Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® Si (25 cm Column)
HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® Si (25 cm Column)
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 30 °C
mobile phase [A] hexane; [B] ethyl acetate
gradient 10 to 30% B in 10 min; held at 30% B for 2 min
flow rate 1.0 mL/min
injection 10 µL
detector UV, 290 nm
Description
Analysis Note Ascentis silica is used in the analysis of fat-soluble vitamins using a normal phase gradient. These include vitamin K 3 (menadione), vitamin E (alpha and gamma tocopherol), and vitamin D3 (cholecalciferol).
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products