Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Fat Soluble Vitamins on Ascentis® Express RP-Amide, Methanol Mobile Phase
HPLC Analysis of Fat Soluble Vitamins on Ascentis® Express RP-Amide, Methanol Mobile Phase
Conditions
column Ascentis Express RP-Amide, 15 cm x 4.6 mm I.D., 2.7 µm particles (53931-U)
column temp. 30 °C
mobile phase methanol
flow rate 1.0 mL/min
pressure 1653 psi (114 bar)
sample 0.4 mg/mL tocopherol acetate, alpha-tocopherol and K1, 0.04 mg/mL D2 and D3, all in ethanol
injection 5 µL
detector UV, 280 nm
Description
Analysis Note Analysis was conducted using standard solution, detection via UV.
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products