HPLC Analysis of Fenthion Sulfoxide Enantiomers on Astec CHIROBIOTIC(R) TAG in Polar Ionic Mode
HPLC Analysis of Fenthion Sulfoxide Enantiomers on Astec CHIROBIOTIC(R) TAG in Polar Ionic Mode
Description
Analysis Note Fenthion sulfoxide is an organothiophosphate insecticide. It comprises two enantiomers by virtue of its sulfur chiral center. Enantioselective influences of chiral pesticides and their metabolites on ecosystems is an area of great importance. The possibility that only one enantiomer has the wanted properties and/or the other form an unwanted effect may exist. An analytical separation of the enantiomers is required to perform studies in these areas. The Astec CHIROBIOTIC® TAG column resolved the enantiomers of fenthion sulfoxide under LC/MS-friendly polar ionic mobile phase conditions.
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC