HPLC Analysis of Fenthion Sulfoxide Enantiomers on Astec(R) CHIROBIOTIC(R) TAG in Reversed Phase Mode
HPLC Analysis of Fenthion Sulfoxide Enantiomers on Astec(R) CHIROBIOTIC(R) TAG in Reversed Phase Mode
Description
Analysis Note Fenthion sulfoxide is an organothiophosphate insecticide. It comprises two enantiomers by virtue of its sulfur chiral center. Enantioselective influences of chiral pesticides and their metabolites on ecosystems is an area of great importance. The possibility that only one enantiomer has the wanted properties and/or the other form an unwanted effect may exist. An analytical separation of the enantiomers is required to perform studies in these areas. The Astec® CHIROBIOTIC® TAG column resolved the enantiomers of fenthion sulfoxide under reversed-phase conditions.
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC