Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Flakka (alpha-PVP) and alpha-Pyrrolidinobutiophenone on Ascentis(R) Express HILIC 5 mum
HPLC Analysis of Flakka (alpha-PVP) and alpha-Pyrrolidinobutiophenone on Ascentis® Express HILIC 5 mum
Conditions
column Ascentis Express HILIC, 5 cm x 3.0 mm I.D., 5 μm particles (50262-U)
column temp. 35 °C
mobile phase 5 mM ammonium formate in 95:5, acetonitrile:water
flow rate 0.8 mL/min
pressure 453 psi (31.2 bar)
sample 50 μg/mL in acetonitrile
injection 2 μL
detector UV, 250 nm
Description
Analysis Note The rapid separation of the street drug Flakka (α-PVP) and related compound is shown here on an Ascentis Express HILIC column. Highest grade UHPLC solvents were used to supply low background interference and low particulate contamination for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Featured Industry Forensics and Toxicology