Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND® I 2000 DMP, IPA:Heptane Mobile Phase
HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND® I 2000 DMP, IPA:Heptane Mobile Phase
Conditions
column CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 µm particles (20724AST)
column temp. 25 °C
mobile phase [A] isopropanol; [B] heptane; (30:70, A:B)
flow rate 0.6 mL/min
sample flavanone, 1 mg/mL in heptane
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Flavors, Fragrances
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC