HPLC Analysis of Flavonoids in Red Wine on Chromolith High Resolution RP-18e (analyte set 1)
HPLC Analysis of Flavonoids in Red Wine on Chromolith High Resolution RP-18e (analyte set 1)
Conditions
column Chromolith High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase 19ml acetonitril / 5ml methanol / 1ml THF / 75ml water
flow rate 2.0 mL/min
pressure 1406 psi 97 Bar
sample Myricetine 907μg/ml, Quercetine 285μg/ml
injection 1 μL
detector 360 nm cell: 11 μL
Description
Featured Industry Food and Beverages