Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Flavonoids in Red Wine on Chromolith High Resolution RP-18e (analyte set 2)
HPLC Analysis of Flavonoids in Red Wine on Chromolith High Resolution RP-18e (analyte set 2)
Conditions
column Chromolith High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase 19ml acetonitril / 5ml methanol / 1ml THF / 75ml water
flow rate 2.0 mL/min
pressure 1406 psi 97 Bar
sample Red wine
injection 1 μL
detector 360 nm (cell: 11 μL)
Description
Featured Industry Food and Beverages