Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Fluoxetine (Prozac®) on Discovery® Cyano
HPLC Analysis of Fluoxetine (Prozac®) on Discovery® Cyano
Conditions
column Discovery Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (59356-U)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] 25 mM KH2PO4, pH 7.0; (40:60, A:B)
flow rate 2 mL/min
injection 1 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC, Prozac is a registered trademark of Eli Lilly and Co.