HPLC Analysis of Furans on Ascentis® Express Phenyl-Hexyl as listed in ASTM method D5837-99
This application demonstrates the suitability of Ascentis Express Hexyl-Phenyl for the efficient separation of furans as listed in ASTM method D5837-99.
HPLC Analysis of Furans on Ascentis® Express Phenyl-Hexyl
Conditions
column Ascentis Express Phenyl-Hexyl, 5 cm x 3.0 mm I.D., 2.7 µm particles (53342-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (80:20, A:B)
flow rate 0.4 mL/min
sample 20 µg/mL in 80:20, water: acetonitrile
injection 1 µL
detector UV, 230 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express Hexyl-Phenyl for the efficient separation of furans as listed in ASTM method D5837-99.
Categories Analytical Chromatography, Furans
Featured Industry Chemicals and Industrial Polymers