Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Furoin on Astec® (R,R) P-CAP™-DP
HPLC Analysis of Furoin on Astec® (R,R) P-CAP™-DP
Conditions
column (R,R) P-CAP-DP, 25 cm x 4.6 mm I.D., 5 µm particles (25024AST)
column temp. 23 °C
mobile phase [A] hexane; [B] ethanol; (80:20, A:B)
flow rate 1.0 mL/min
sample 4 µg/mL
injection 0.2 mL
detector UV, 235 nm
Description
Categories Analytical Chromatography, Flavors, Fragrances
Featured Industry Chemicals and Industrial Polymers
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC