Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Glucocorticoids on Ascentis® C8
HPLC Analysis of Glucocorticoids on Ascentis® C8
Conditions
column Ascentis C8, 15 cm x 4.6 mm I.D., 5 µm particles (581424-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (68:32, A:B)
flow rate 1.0 mL/min
sample as indicated in mobile phase
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, Steroids, hormones, steroids
Featured Industry Clinical