HPLC Analysis of Glyocoalkaloids - Solanine and Cacoline on Chromolith(R) NH2 100 - 4.6mm
HPLC Analysis of Glyocoalkaloids - Solanine and Cacoline on Chromolith® NH2 100 - 4.6mm
Conditions
column Chromolith® NH2, 100 x 4.6 mm (1.52028.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] phosphate buffer, pH 6; (80:20, A:B)
flow rate 2.0 mL/min
pressure 174 psi (12 bar)
sample 200 ppm (0.2 mg/mL) of each Solanine and Cacoline in mobile phase
injection 8 μL
detector 208 nm
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany