Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Heroin, Cocaine, Amphetamine and MDMA on Ascentis® Express HILIC
HPLC Analysis of Heroin, Cocaine, Amphetamine and MDMA on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 50 °C
mobile phase [A] 10 mM ammonium acetate in water, pH 3.5; [B] 10 mM ammonium acetate in acetonitrile; (2.98, A:B)
flow rate 0.4 mL/min
injection 1 µL
detector ESI(+), m/z 100-400
Description
Categories Analytical Chromatography, Forensics and Toxicology, Drugs of Abuse, drugs of abuse
Featured Industry Forensics and Toxicology