Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Hippuric and Methylhippuric Acids in Urine on SUPELCOSIL™ LC-18
HPLC Analysis of Hippuric and Methylhippuric Acids in Urine on SUPELCOSIL™ LC-18
Conditions
column SUPELCOSIL LC-18, 7.5 cm x 4.6 mm I.D., 3 µm particles (58984)
mobile phase [A] tetrahydrofuran:10 mM potassium phosphate, pH 3.0 [B] with phosphoric acid (3:97, A:B)
flow rate 1.5 mL/min
injection 10 µL spiked urine (300 µg/mL HA, o-MHA; 150 µg/mL m-MHA, p-MHA)
detector UV, 260 nm
Description
Analysis Note Urinary excretion of hippuric acid and m-or p-methylhippuric acid in the urine is a test of exposure to toluene and m-or p-xylene vapors.
Categories Analytical Chromatography, Clinical, Forensics and Toxicology, organic acids, Organic Acids
Featured Industry Clinical
Forensics and Toxicology
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC