HPLC Analysis of Hydroxymethylfurfural on Chromolith® High Resolution RP18e
HPLC Analysis of Hydroxymethylfurfural on Chromolith® High Resolution RP18e
Conditions
column Chromolith® High Resolution RP18 endcapped, 100 x 4.6 mm (1.52022.0001)
mobile phase [A] acetonitrile; [B] water ; (10:90, A:B)
flow rate 1.0 mL/min
pressure 900 psi (62 bar)
column temp. ambient
detector 277 nm cell: 11 μL
injection 5 μL
Description
Categories Analytical/Chromatography, Applications, HPLC Applications, Natural products
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany