HPLC Analysis of Ilvico on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Ilvico on Chromolith® High Resolution RP-18e
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase [A] methanol; [B] water + 0,1%TFA; (75:25, A:B)
flow rate 2.0 mL/min
sample Maleic acid 48μg/ml, Paracetamol 42μg/ml, Caffein 49μg/ml, Brompheniramine 30μg/ml
injection 3 μL
detector 220 nm
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany