Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Insulin on Chromolith Performance 100-2mm
HPLC Analysis of Insulin on Chromolith Performance 100-2mm
Conditions
sample preparation 9mg Insulin are dissolved in 9ml 0,01M HCl and put into the vaccuumdryer at 50°C for 4 hours. Then it was stored 96h by 25°C.
column Chromolith® Performance 100 x 2 mm (1.52006.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] Water + 0,1% TFA v/v; (30:70, A:B)
flow rate 95 μL/min
pressure 1581 psi (109 bar)
sample Insulin 1mg/mL in 10mM HCl
injection 0.4 μL
detector 215 nm cell: 1.4 μL