Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Insulin (Species Variants) on Discovery® BIO Wide Pore C8
HPLC Analysis of Insulin (Species Variants) on Discovery® BIO Wide Pore C8
Conditions
column Discovery BIO Wide Pore C8, 15 cm x 4.6 mm I.D., 5 µm particles (568322-U)
column temp. 30 °C
mobile phase [A] (0.1% TFA in water):(0.1% TFA in acetonitrile), 71:29 [B]: (0.1% TFA in water):(0.1% TFA in acetonitrile), 68:32,
gradient 0-100% B in 30 min
flow rate 1 mL/min
injection 5 µLin 0.1% TFA containing 5ug each of three insulin variants
detector UV, 215 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC