HPLC Analysis of Isocyanates on Titan™ C18
HPLC Analysis of Isocyanates on Titan™ C18
column Titan C18, 5 cm x 2.1 mm I.D., 1.9 µm particles (577122-U)
column temp. 30 °C
mobile phase [A] 0.05% formic acid in 5:95 acetonitrile:water; [B] 0.05% formic acid in 95:5 acetonitrile:water
gradient 50% to 90% B in 1 min, hold at 90% B for 3 min, 90% to 50% B in 0.1 min, hold at 50% B for 3.9 min
flow rate 0.5 mL/min
pressure 3916 psi (270 bar)
sample isocyanates in acetonitrile, 0.06 µg/mL
injection 2 µL
detector MS, ESI (+)
detector MRM
Analysis Note Analysis of DBA-derivatized isocyanate monomers on Titan C18 using LC-MS/MS.
Categories Analytical Chromatography, Air Monitoring Applications, Isocyanates
Featured Industry Industrial Hygiene
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC