Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Isoproterenol Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Isoproterenol Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] methanol; [C] acetic acid; [D] triethylamine; (55:45:0.3:0.2, A:B:C:D)
flow rate 2 mL/min
sample 5 mg/mL in methanol
injection 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Bronchodilators, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC