HPLC Analysis of Itraconazol on Chromolith(R) Performance 100 - 4.6mm
HPLC Analysis of Itraconazol on Chromolith® Performance 100 - 4.6mm
sample preparation 50mg Itraconazol dissolved in 50mL Solution (5mL HCl 30% in 1L Methanol) to get the impurities.
column Chromolith® Performance 100 x 4.6 mm (1.52028.0001)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] methanol; [C] 1L Water + 3,4g TBAH-sulfat; (45:10:45, A:B:C)
flow rate 2.0 mL/min
pressure 4025 psi (28 bar)
sample Itraconazol 1mg/1mL Solution
injection 10 μL
detector 225 nm cell: 11 μL
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany