HPLC Analysis of Ivermectin Injection as per USP/BP on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Ivermectin Injection as per USP/BP on Chromolith® High Resolution RP-18e
Conditions
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] methanol: [C] water; (106:55:39, A:B:C)
flow rate 1.5 mL/min
sample 0.4 mg / ml in diluent
injection 10 μL
detector 245 nm cell: 10 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany