HPLC Analysis of β-Lactam Antibiotics on Discovery® RP-Amide C16
column Discovery RP-Amide C16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 30 °C
mobile phase [A] 0.01% TFA in water; [B] 0.01% TFA in acetonitrile
flow rate 1.5 mL/min
detector UV, 254 nm
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics, Antibiotics, Antimicrobials
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC