HPLC Analysis of Levetiracetam on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Levetiracetam on Chromolith® High Resolution RP-18e
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (90:10, A:B)
flow rate 1 mL/min
sample Levetiracetam 50μg/ml, Caffein 50μg/ml
injection 10 μL
detector 200 nm (cell: 11 μL)
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany