HPLC Analysis of Levetiracetam on Chromolith(R) Performance 100 - 4.6 mm
HPLC Analysis of Levetiracetam on Chromolith® Performance 100 - 4.6 mm
Conditions
column 1.52028.0001 (1.52028)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (90:10, A:B)
flow rate 1 mL/min
pressure 6613 psi (456 bar)
sample Levetiracetam 50μg/ml, Caffein 50μg/ml
injection 10 μL
detector 200 nm cell: 11 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability suitable for HPLC