HPLC Analysis of Levetiracetam on Chromolith(R) Performance 100-3mm
HPLC Analysis of Levetiracetam on Chromolith® Performance 100-3mm
column 1.52001.0001 (1.52001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (90:10, A:B)
flow rate 425 μL/min
pressure 4169 psi (287 bar)
sample Levetiracetam 50μg/ml, Caffein 50μg/ml
injection 5 μL
detector 200 nm cell: 1.4 μL
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany