HPLC Analysis of Levetiracetam on Chromolith(R) Performance 100- 2 mm
HPLC Analysis of Levetiracetam on Chromolith® Performance 100- 2 mm
Conditions
column Chromolith® Performance 100 x 2 mm (1.52006.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (90:10, A:B)
flow rate 190 μL/min
pressure 450 psi (31bar)
sample Levetiracetam 50μg/ml, Caffein 50μg/ml
injection 2.0 μL
detector 200 nm cell: 1.4 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany