HPLC Analysis of Linear Oligosaccharides on Chromolith(R) NH2 100 - 4.6mm
HPLC Analysis of Linear Oligosaccharides on Chromolith® NH2 100 - 4.6mm
Conditions
column Chromolith® NH2 100 x 4.6 mm (1.52028.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] water; (65:35, A:B)
flow rate 1.0 mL/min
pressure 377 psi (26 bar)
sample Maltose 21.6 mg/mL. Maltotriose 19.5 mg/mL, Maltotetraose 15.7 mg/mL, Maltopentaose 16.2 mg/mL, Maltoheptaose 10.6 mg/mL in mobile phase
injection 2 μL
detector 190 nm
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany