HPLC Analysis of Loratadine on Chromolith(R) HighResolution RP-18 endcapped
HPLC Analysis of Loratadine on Chromolith® HighResolution RP-18 endcapped
Conditions
column Purospher® STAR RP-18 endcapped 150 x 4.6 mm (1.52022.0001)
column temp. 30 °C
mobile phase [A] 2.28g K2HPO4 in 800mL H2O, adjust pH 6.0 with H3PO4, add H2O to 1000mL; [B] methanol; (20:80, A:B)
flow rate 1 mL/min
pressure 899 psi (62 Bar)
sample Weigh appropriate amount of Loratadine to obtain 200 μg/mL (200 ppm). Dissolve with diluent.
injection 10 μL
detector 247 nm (cell: 1 μL/10 mm)
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany