HPLC Analysis of Lutein on Ascentis® Express C18
HPLC Analysis of Lutein on Ascentis® Express C18
sample preparation SPE (Solid Phase Extraction)
column Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 mum particles (53822-U)
column temp. 30 °C
mobile phase [A] Acetonitrile; [B] Water; (90:10, A:B)
flow rate 1.0 mL/min
pressure 2581 psi (178 bar)
sample 0.10 mg/mL of Lutein in 50/50 MeOH/THF
injection 0.3 µL
detector UV, 450 nm
Analysis Note The analysis of lutein is relevant to pharmaceutical, nutraceutical, food, pet foods, and animal and fish feed applications. This method uses a 2.7 μm Ascentis® Express C18 column for a rapid analysis of lutein in under two minutes with UV detection.
Categories Analytical Chromatography, Pigments
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Pharmaceutical (small molecule)
Vitamins, Nutraceuticals, and Natural Products