Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Melamine in Infant Forumula on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Melamine in Infant Forumula on Chromolith® High Resolution RP-18e
Conditions
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. 40 °C
mobile phase [A] acetonitrile; [B] pH=3 (10mM citric acid, 10mM sodium octan sulfonate; (15:85, A:B)
flow rate 0.8 mL/min
sample 10 mL infant formula spiked with 25 μg melamine
injection 2 μL
detector 240 nm cell: 11 μL
Description
Analysis Note The injected sample is a 25 μg melamine spiked to 10 mL milk ans diluted with 1 mL acetic acid 3% v/v and stored at 5 °C for 30 min. After 15 min of centrifugation (10,000 rpm) supernatants were filtered through a 0.45 μm Millex PVDF filter and injected.
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany