HPLC Analysis of Methacrylamide, Acrylamide, and Methacrylic Acid on SeQuant® ZIC-HILIC
HPLC Analysis of Methacrylamide, Acrylamide, and Methacrylic Acid on SeQuant ZIC-HILIC
Conditions
column SeQuant® ZIC-HILIC, PEEK 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase 95% Acetonitrile and 5% Acetic Acid 50 mM (v/v)
flow rate 1.0 mL/min
pressure 8.0 MPa
detection UV @ 210 nm (UFS 1.0 V)
temperature Ambient
inject. volume 5.0 μL
sample Methacrylamide, Acrylamide, Methacrylic Acid standards diluted in mobile phase
Description
Featured Industry Chemicals and Industrial Polymers